Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SuperHM-L HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021916 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1916 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1916 Supplier MilliporeSigma Supelco Supplier No. 817998
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SuperHM-L Mixed-Bed Columns are packed with rigid polymer beads in varying pore sizes. They are used for the non-aqueous separation of polymers with a broad molecular weight range, or for analyzing unknown samples.

  • Columns are shipped in THF, which can be exchanged for a wide variety of organic solvents
  • High efficiency/high throughput versions of conventional TSKgel HHR Columns
  • Both column types are based on the same bead chemistry
  • High-throughput gel permeation columns have dimensions of 6 mm ID x 15 cm
  • Maximum Operating Temperature: 140°C
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 6 mm
Flow Rate 0.8 mL/min
Length (Metric) 15 cm
Max. Pressure 39 bar
Particle Size 3 μm
Pore Size Mixed
Temperature 140°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Matrix Polystyrene-divinylbenzene Phase/Polymer Particle Platform
Phase Size Exclusion
Material Stainless Steel Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.