Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SuperHZM-H Guard Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022021 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2 cm
3.5 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-2021 3.5 cm
11-102-2022 2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2021 Supplier MilliporeSigma Supelco Supplier No. 819667
Add to Cart
Add to Cart

TSKgel™ SuperHZM-H Mixed-Bed Columns are packed with rigid, porous, polymer beads each having a varying mean pore diameter. They are used for the non-aqueous separation of organic-soluble polymers with high molecular weights of less than 400,000,000 Da.

  • THF shipping solvent may be replaced by a limited number of organic solvents
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 10 μm
Length (Metric) 3.5 cm
Diameter (Metric) Inner 4.6 mm
Flow Rate 0.4 mL/min
Stationary Phase Polystyrene-divinylbenzene
Temperature (Metric) Maximum 80°C
Material Stainless Steel
Technique Size Exclusion (SEC)
Product Line TSKgel

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.