Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SuperOligoPW HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022177 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-2177 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2177 Supplier MilliporeSigma Supelco Supplier No. 822792
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TSKgel™ SuperOligoPW column is designed for the determination of molecular mass of aqueous oligomers, particularly oligosaccharides, and low molecular weight aqueous polymers.

Compared to the TSKgel™ G-Oligo-PW column, the three micron particle size and semi-micro dimensions (15 cm x 6 mm I.D.) of the TSKgel™ SuperOligoPW column reduces analysis time by a factor of two while maintaining resolution and lowering solvent consumption.Since the packing material in the TSKgel™ SuperOligoPW columns is more hydrophilic compared with TSKgel™ G-Oligo-PW columns, an even wider range of water-soluble polymers can be eluted without the need to add organic solvent to the eluent.

Order Info

Shipped in water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 6 mm
Flow Rate 0.6 mL/min.
Length (Metric) 15 cm
Max. Pressure 50 bar
Particle Size 3 μm
pH 2.0 - 12
Pore Size 125 Å
Solvents ≤20% polar organic solvent
Temperature 10°C to 80°C
USP Type L38, L39
Stationary Phase Hydroxylated Methacrylate
Product Line TSKgel
Matrix Hydroxylated Methacrylic Polymer
Phase Size Exclusion
Material Stainless Steel
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.