Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SWXL Column, 7 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021922 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
4 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1922 4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1922 Supplier MilliporeSigma Supelco Supplier No. 818008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel SWXL Columns feature highly porous silica particles, the surface of which has been shielded from interacting with proteins by derivatization with ligands containing diol functional groups.

  • TSKgel SWXL-type columns contain smaller particles than TSKgel SW-type columns
  • Large pore volumes allow TSKgel SWXL-type columns to stand out from other silica- or polymer-based high performance size exclusion columns
  • These columns are commonly used in the quality control of monoclonal antibodies and other biopharma products
  • Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7
  • Salt Concentration: ≤ 0.5 M
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 6 mm
Flow Rate 1.2 mL/min
Length (Metric) 4 cm
Particle Size 7 μm
pH 2.5 to 7.5
Pore Size 25 Å
Solvents 0-100% Aqueous Soluble Organic Solvents
Temperature 10°C to 30°C
USP Type L20, L59
Stationary Phase Diol
Product Line TSKgel
Shipping Condition Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer
Matrix Diol Phase/Silica Particle Platform
Material PEEK Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.