Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SWXL Column, 8 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022040 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-2040 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2040 Supplier MilliporeSigma Supelco Supplier No. 820025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An excellent choice for pegylated proteins or glycosylated biomolecules. The fractionation range is applicable for globular protein samples up to 7 million Da.

  • Made of PEEK housing material to reduce sample absorption to stainless steel or glass
  • These columns have the same attributes as the corresponding stainless steel columns (TSKgel G2000SWxl, G3000Swxl and G4000SWxl)
  • Widely considered the premium columns for analyzing biopolymers by GFC
  • TSKgel G4000SWxl columns feature high pore volume per unit column volume, low sample adsorption, and excellent column efficiency all contributing to unsurpassed sample resolution
  • With 8 micron particles and 450 Å pores, TSKgel G4000SWxl provides the largest exclusion limit and the widest fractionation range
  • Calibration Range: 20,000 to 7 x 106 Da (globular proteins)
  • Salt Concentration: ≤ 0.5 M
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1.2 mL/min
Length (Metric) 30 cm
Max. Pressure 72 bar
Particle Size 8 μm
pH 2.5 to 7.5
Pore Size 450 Å
Solvents 0-100% Aqueous Soluble Organic Solvents
Temperature 10°C to 30°C
USP Type L20, L59
Stationary Phase Diol
Product Line TSKgel
Shipping Condition Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer, pH 6.7
Matrix Diol Phase/Silica Particle Platform
Material PEEK Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.