Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ UltraSW Aggregate HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022196 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-2196 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2196 Supplier MilliporeSigma Supelco Supplier No. 822856
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ UltraSW Aggregate columns have a slightly larger pore size and smaller (3 μm) particle size than the industry-leading TSKgel™ G3000SWxl (5 μm) and SuperSW3000 (4 μm) columns. These 30 cm x 7.8 mm I.D. silica-based, stainless steel columns offer reduced lot-to-lot variation, long column life, reduction of unspecified adsorption, and superior recovery of aggregates.

The calibration curve of the TSKgel™ UltraSW Aggregrate column shows a separation range up to around 2 million Da, which provides better resolution of aggregates/multimers of a mAb than the TSKgel™ SuperSW mAb columns.

Order Info

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1 mL/min.
Length (Metric) 30 cm
Max. Pressure 120 bar
Particle Size 3 μm
pH 2.5 - 7.5
Pore Size 300 Å
Solvents 0 to 100% aqueous soluble organic solvents
Temperature 10°C to 30°C
USP Type L20, L33, L59
Stationary Phase Diol
Product Line TSKgel
Matrix Silica Particle Platform
Phase Size Exclusion
Material Stainless Steel
Technique SEC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.