Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSKS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544118 Supplier Invitrogen™ Supplier No. PA544118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MASVVVKTIW QSKEIHEAGD TPTGVESCSQ LVPEAPRRVT SRAKGIPKKK Sequence homology: Cow: 86%; Dog: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 79%; Rat: 86%.

A novel putative serine/threonine kinase substrate gene (TSKS, for testis-specific kinase substrate) is likely the human homolog of the mouse testis-specific serine kinase substrate gene. TSKS exhibits high levels of expression exclusively in human testicular tissue. The expression of TSKS is downregulated in cancerous testicular tissue, in comparison to adjacent normal tissue. These observations suggest a role of TSKS in testicular physiology, most probably in the process of spermatogenesis or spermiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSKS
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TSKS
Gene Accession No. Q9UJT2
Gene Alias clone 4; PPP1R161; protein phosphatase 1, regulatory subunit 161; serine/threonine kinase 22 substrate 1; STK22 substrate 1; STK22S1; testis specific serine kinase substrate; testis specific serine/threonine kinase substrate; testis-specific kinase substrate; Testis-specific serine kinase substrate; testis-specific serine/threonine kinase substrate 1; TSKS; TSKS1; TSSKS; Tssks1; Unknown (protein for MGC:134104)
Gene Symbols TSKS
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TSKS (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60385
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.