Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKS Polyclonal Antibody, Invitrogen™

Catalog No. PIPA530961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530961 Supplier Thermo Scientific Supplier No. PA530961

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (90%), Rat (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

A novel putative serine/threonine kinase substrate gene (TSKS, for testis-specific kinase substrate) is likely the human homolog of the mouse testis-specific serine kinase substrate gene. TSKS exhibits high levels of expression exclusively in human testicular tissue. The expression of TSKS is downregulated in cancerous testicular tissue, in comparison to adjacent normal tissue. These observations suggest a role of TSKS in testicular physiology, most probably in the process of spermatogenesis or spermiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSKS
Applications Western Blot
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TSKS
Gene Accession No. Q9UJT2
Gene Alias clone 4; PPP1R161; protein phosphatase 1, regulatory subunit 161; serine/threonine kinase 22 substrate 1; STK22 substrate 1; STK22S1; testis specific serine kinase substrate; testis specific serine/threonine kinase substrate; testis-specific kinase substrate; Testis-specific serine kinase substrate; testis-specific serine/threonine kinase substrate 1; TSKS; TSKS1; TSSKS; Tssks1; Unknown (protein for MGC:134104)
Gene Symbols TSKS
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 282 and 574 of Human TSKS
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60385
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.