Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSKU Monoclonal Antibody (462310)
GREENER_CHOICE

Catalog No. PIMA524182
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524182 Supplier Invitrogen™ Supplier No. MA524182
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human Tsukushi/TSK is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

The TSKU gene encodes Tsukushi, a member of the small leucine-rich proteoglycan (SLRP) family, playing crucial roles in cellular signaling, growth, and development. Tsukushi is characterized by its involvement in various biological processes, including immune regulation, tissue remodeling, and metabolic homeostasis. It is a hepatokine released in response to conditions like nonalcoholic fatty liver disease (NAFLD), influencing metabolic traits and contributing to conditions such as dyslipidemia and cardiovascular diseases. Tsukushi impacts energy expenditure by modulating brown fat thermogenesis, thereby affecting metabolic homeostasis and obesity-related metabolic traits. As an extracellular matrix component, Tsukushi participates in signaling pathways that regulate cell proliferation, migration, and differentiation, impacting organ development and pathophysiological processes. Its role in metabolic and inflammatory diseases presents it as a potential biomarker and therapeutic target in conditions characterized by altered metabolic and inflammatory states. Ongoing research further elucidates Tsukushi's mechanisms and interactions, contributing to our understanding of its multifunctional roles in health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSKU
Applications Western Blot
Classification Monoclonal
Clone 462310
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TSKU
Gene Accession No. Q8CBR6
Gene Alias 9530051K01Rik; E2ig4; E2-induced gene 4 protein; Early insulin-induced hepatic gene protein; Eiih; hepatic protein EIIH; leucine rich repeat containing 54; leucine-rich repeat-containing protein 54; LRRC54; Tsk; Tsku; Tsukushi; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushi, small leucine rich proteoglycan; tsukushin; UNQ850/PRO1788
Gene Symbols TSKU
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Tsukushi/TSK Thr18-Leu354.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244152
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.