Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSLP Antibody (55N1D10), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227326C
Description
TSLP Monoclonal specifically detects TSLP in Human, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TSLP | |
Monoclonal | |
DyLight 650 | |
Q969D9 | |
TSLP | |
A partial recombinant TSLP protein (amino acids 27-158) was used as the immunogen for the antibody. | |
0.1 mL | |
Primary | |
Human, Rabbit | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
55N1D10 | |
Western Blot, Immunohistochemistry-Paraffin | |
thymic stromal lymphopoietin | |
Mouse | |
Protein G purified | |
RUO | |
85480 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction