Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSLP Monoclonal Antibody (152640)

Catalog No. PIMA523779
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523779 500 μg
PIMA523875 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA523779 Supplier Invitrogen™ Supplier No. MA523779

Rat Monoclonal Antibody

In Western blots, this antibody does not cross-react with recombinant mouse IL-7 or recombinant human TSLP. Reconstitute at 0.5 mg/mL in sterile PBS.

TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSLP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 152640
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TSLP
Gene Accession No. Q9JIE6
Gene Alias Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP
Gene Symbols TSLP
Host Species Rat
Immunogen Sf 21-derived recombinant mouse TSLP Tyr20-Glu140.
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53603
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.