Learn More
Invitrogen™ TSLP Polyclonal Antibody

Description
Despite its predicted molecular weight, TSLP often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is Jurkat cell lysate. PA5-20319 can be used with blocking peptide PEP-0439.
Specifications
Specifications
| Antigen | TSLP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TSLP |
| Gene Accession No. | Q969D9 |
| Gene Alias | Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP |
| Gene Symbols | TSLP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.