Learn More
TSLP Polyclonal Antibody, Biotin, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $638.37
Specifications
| Antigen | TSLP |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1439
![]()
|
Rockland Immunochemicals
209406H48 |
100 μg |
Each for $638.37
|
|
|||||
50-199-1440
![]()
|
Rockland Immunochemicals
209406H48S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-TSLP is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. This antibody is specific for human TSLP protein. A BLAST analysis was used to suggest cross-reactivity with TSLP from human sources based on 100% homology with the immunizing sequence. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin and anti-Rabbit Serum. Cross-reactivity with TSLP from other sources has not been determined.
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.Specifications
| TSLP | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| Q969D9 | |
| 85480 | |
| Anti-TSLP antibody was prepared from rabbit antiserum after repeated immunizations with a truncated form of recombinant human TSLP protein from E. coli. | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TSLP |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
| Thymic stroma-derived lymphopoietin; Thymic stromal lymphopoietin; TSLP | |
| TSLP | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.