Learn More
TSLP Receptor Monoclonal Antibody (eBio1A6 (1A6)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$1562.00
Specifications
Antigen | TSLP Receptor |
---|---|
Clone | eBio1A6 (1A6) |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-549-937
![]() |
LIFE TECHNOLOGIES
14549937 |
2 mg |
Each of 1 for $1,562.00
|
|
|||||
Description
The eBio1A6 MAb reacts with human thymic stromal-derived lymphopoietin receptor (TSLPR). TSLPR is an approximately 50 kDa protein with significant similarity to the common gamma-chain. TSLPR complexes with IL-7R alpha (CD127) to form the high affinity receptor that binds thymic stromal-derived lymphopoietin (TSLP). Human TSLPR is expressed by monocytes and CD11c+ dendritic cells, and TSLP binding induces the expression of the Th2 cell-attracting chemokines CCL17 and CCL22. Furthermore, the TSLPR-induced activation of dendritic cells indirectly results in the increased secretion of Th cytokines IL-4, -5 and -13, which may be necessary for the regulation of CD4+ T cell homeostasis. In mice, deficiency of TSLPR has no effect on lymphocyte numbers, whereas double deficiency of TSLPR and common gamma-chain results in fewer lymphocytes than seen in mice deficient in the common gamma-chain alone. The eBio1A6 MAb is able to cross-block binding of another anti-human TSLPR MAb, 1D8.
Thymic stromal lymphopoietin (TSLP) has recently been identified as an important factor capable of driving dendritic cell maturation and activation. It is involved in the positive selection of regulatory T cells, maintenance of peripheral CD4+ T cell homeostasis and the induction of CD4+ T cell-mediated allergic reaction. TSLP is also capable of supporting the growth of fetal liver and adult B cell progenitors and their differentiation to the IgM-positive stage of B cell development. Its receptor TSLP-R will bind TSLP in a low-affinity fashion in transfected cells; co-transfection with IL-7R-alpha cDNA results in high-affinity binding and a functional heteromeric complex. This heteromeric receptor requires stat5 for TSLP-mediated signal transduction and is inhibited by SOCS-1.Specifications
TSLP Receptor | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
64109 | |
Primary | |
4°C | |
CRLF2 |
eBio1A6 (1A6) | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9HC73 | |
IgG2a λ | |
Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.