Learn More
Invitrogen™ TSLP Receptor Monoclonal Antibody (eBio1A6 (1A6)), PE, eBioscience™
Mouse Monoclonal Antibody
$411.00
Specifications
Antigen | TSLP Receptor |
---|---|
Clone | eBio1A6 (1A6) |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Description
Description: The eBio1A6 monoclonal antibody reacts with human thymic stromal-derived lymphopoietin receptor (TSLPR). TSLPR is an approximately 50 kDa protein with significant similarity to the common gamma-chain. TSLPR complexes with IL-7R alpha (CD127) to form the high affinity receptor that binds thymic stromal-derived lymphopoietin (TSLP). Human TSLPR is expressed by monocytes and CD11c+ dendritic cells, and TSLP binding induces the expression of the Th2 cell-attracting chemokines CCL17 and CCL22. Furthermore, the TSLPR-induced activation of dendritic cells indirectly results in the increased secretion of Th cytokines IL-4, -5 and -13, which may be necessary for the regulation of CD4+ T cell homeostasis. In mice, deficiency of TSLPR has no effect on lymphocyte numbers, whereas double deficiency of TSLPR and common gamma-chain results in fewer lymphocytes than seen in mice deficient in the common gamma-chain alone. The eBio1A6 monoclonal antibody is able to cross-block binding of another anti-human TSLPR monoclonal antibody, 1D8. Applications Reported: This eBio1A6 (1A6) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio1A6 (1A6) antibody has been pre-titrated and tested by flow cytometric analysis of human TSLPR-transfected Ba/F3 cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be deter...
Thymic stromal lymphopoietin (TSLP) has recently been identified as an important factor capable of driving dendritic cell maturation and activation. It is involved in the positive selection of regulatory T cells, maintenance of peripheral CD4+ T cell homeostasis and the induction of CD4+ T cell-mediated allergic reaction. TSLP is also capable of supporting the growth of fetal liver and adult B cell progenitors and their differentiation to the IgM-positive stage of B cell development. Its receptor TSLP-R will bind TSLP in a low-affinity fashion in transfected cells; co-transfection with IL-7R-alpha cDNA results in high-affinity binding and a functional heteromeric complex. This heteromeric receptor requires stat5 for TSLP-mediated signal transduction and is inhibited by SOCS-1.Specifications
TSLP Receptor | |
5 μL/Test | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q9HC73 | |
64109 | |
Primary | |
Affinity chromatography |
eBio1A6 (1A6) | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG2a λ | |
Human | |
CRLF2 | |
CRL2; Crlf2; CRLF2Y; Crlm2; CRLM-2; cytokine receptor CRL2 precusor; Cytokine receptor-like 2; Cytokine receptor-like factor 2; cytokine receptor-like molecule 2; delta 1; ILXR; IL-XR; Ly114; lymphocyte antigen 114; P2RY8/CRLF2 fusion; thymic stromal lymphopoietin protein receptor; thymic stromal-derived lymphopoietin receptor; thymic stromal-derived lymphopoietin, receptor; Tpte2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2; TSLP receptor; Tslpr; type I cytokine receptor delta 1; WI2-87105E3.1 | |
CRLF2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.