Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSLP Receptor Rat anti-Mouse , PE, Clone: 22H9, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566673
Description
TSLP Receptor Monoclonal antibody specifically detects TSLP Receptor in Mouse samples. It is validated for Flow Cytometry
Specifications
TSLP Receptor | |
Monoclonal | |
PE | |
TSLP receptor; Tslpr; Ly114; Crlf2; CRLM2; Tpte2 | |
TSLP Receptor Protein | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
22H9 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction