Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSLPR/CRLF2 Antibody (59N5G4), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227331V
Description
TSLPR/CRLF2 Monoclonal specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TSLPR/CRLF2 | |
Monoclonal | |
DyLight 405 | |
Q9HC73 | |
CRLF2 | |
Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody. | |
0.1 mL | |
Primary | |
TSLPR is a glycoprotein with two major isoforms of 359 amino acids and 255 amino acids. In western blotting, bands may be visualized between 27 kD and ∼50 kD. | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry (Paraffin) | |
59N5G4 | |
Western Blot, Immunohistochemistry-Paraffin | |
CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR | |
Mouse | |
Protein G purified | |
RUO | |
64109 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction