Learn More
TSN Mouse anti-Human, Clone: OTI4C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809193

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Translin is a DNA-binding protein which specifically recognizes conserved target sequences at the breakpoint junction of chromosomal translocations. Translin polypeptides form a multimeric structure that is responsible for its DNA-binding activity. Recombination-associated motifs and translin-binding sites are present at recombination hotspots and may serve as indicators of breakpoints in genes which are fused by translocations. These binding activities may play a crucial role in chromosomal translocation in lymphoid neoplasms. Recent evidence suggests that in combination with a binding partner Trax, Translin binds RNA and mediates trafficking in neurons.Specifications
TSN | |
Monoclonal | |
Unconjugated | |
TSN | |
BCLF-1, C3PO, RCHF1, REHF-1, TBRBP, TRSLN | |
Mouse | |
Affinity Chromatography | |
RUO | |
7247 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15631 | |
TSN | |
Human recombit protein fragment corresponding to amino acids 1-51 of human TSN produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.