Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521150
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521150 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521150 Supplier Invitrogen™ Supplier No. PA521150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat lung tissue lysate. PA5-21150 can be used with blocking peptide PEP-1264.

Translin is a DNA-binding protein which specifically recognizes conserved target sequences at the breakpoint junction of chromosomal translocations. Translin polypeptides form a multimeric structure that is responsible for its DNA-binding activity. Recombination-associated motifs and translin-binding sites are present at recombination hotspots and may serve as indicators of breakpoints in genes which are fused by translocations. These binding activities may play a crucial role in chromosomal translocation in lymphoid neoplasms. Recent evidence suggests that in combination with a binding partner Trax, Translin binds RNA and mediates trafficking in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSN
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Tsn
Gene Accession No. Q15631, Q62348
Gene Alias 2610034C24Rik; AU040286; BCLF-1; C3PO; Component 3 of promoter of RISC; RCHF1; recombination hotspot associated factor; recombination hotspot-binding protein; REHF-1; TBRBP; TB-RBP; testis brain-RNA binding protein; testis/brain RNA-binding protein; testis/brain-RNA-binding protein; translin; TRSLN; Tsn
Gene Symbols Tsn
Host Species Rabbit
Immunogen A 20 amino acid peptide near the carboxy terminus of human Translin.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22099, 60381, 7247
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.