Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116869 Supplier Invitrogen™ Supplier No. PA5116869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: K-562, HeLa, Mouse spleen, Mouse brain, Rat thymus, Rat spleen Immunogen sequence: GFQDIPKRCL KAREHFGTVK THLTSLKTKF PAEQYYRFHE HWRFVLQRLV FLAAFVVYLE TETLVTREAV TEILGIEPDR EKGFHLDVED YLSGVLILAS E.

Translin is a DNA-binding protein which specifically recognizes conserved target sequences at the breakpoint junction of chromosomal translocations. Translin polypeptides form a multimeric structure that is responsible for its DNA-binding activity. Recombination-associated motifs and translin-binding sites are present at recombination hotspots and may serve as indicators of breakpoints in genes which are fused by translocations. These binding activities may play a crucial role in chromosomal translocation in lymphoid neoplasms. Recent evidence suggests that in combination with a binding partner Trax, Translin binds RNA and mediates trafficking in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Tsn
Gene Accession No. Q15631, Q62348
Gene Alias 2610034C24Rik; AU040286; BCLF-1; C3PO; Component 3 of promoter of RISC; RCHF1; recombination hotspot associated factor; recombination hotspot-binding protein; REHF-1; TBRBP; TB-RBP; testis brain-RNA binding protein; testis/brain RNA-binding protein; testis/brain-RNA-binding protein; translin; TRSLN; Tsn
Gene Symbols Tsn
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22099, 60381, 7247
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.