Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563747
Description
Immunogen sequence: MSVSEIFVEL QGFLAAEQDI REEIRKVVQS LEQTAREILT LLQGVHQGAG F Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Translin is a DNA-binding protein which specifically recognizes conserved target sequences at the breakpoint junction of chromosomal translocations. Translin polypeptides form a multimeric structure that is responsible for its DNA-binding activity. Recombination-associated motifs and translin-binding sites are present at recombination hotspots and may serve as indicators of breakpoints in genes which are fused by translocations. These binding activities may play a crucial role in chromosomal translocation in lymphoid neoplasms. Recent evidence suggests that in combination with a binding partner Trax, Translin binds RNA and mediates trafficking in neurons.
Specifications
TSN | |
Polyclonal | |
Unconjugated | |
Tsn | |
2610034C24Rik; AU040286; BCLF-1; C3PO; Component 3 of promoter of RISC; RCHF1; recombination hotspot associated factor; recombination hotspot-binding protein; REHF-1; TBRBP; TB-RBP; testis brain-RNA binding protein; testis/brain RNA-binding protein; testis/brain-RNA-binding protein; translin; TRSLN; Tsn | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7247 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15631 | |
Tsn | |
Recombinant protein corresponding to Human TSN. Recombinant protein control fragment (Product #RP-101621). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction