Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSNAX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556625 Supplier Invitrogen™ Supplier No. PA556625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IFQVAQELSG EDMHQFHRAI TTGLQEYVEA VSFQHFIKTR SLISMDEINK QLIFTTEDNG KENKTPSSDA QDKQFGTWRL RVTPVDYL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

This gene encodes a protein which specifically interacts with translin, a DNA-binding protein that binds consensus sequences at breakpoint junctions of chromosomal translocations. The encoded protein contains bipartite nuclear targeting sequences that may provide nuclear transport for translin, which lacks any nuclear targeting motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSNAX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSNAX
Gene Accession No. Q99598
Gene Alias R74833; translin associated factor X; translin-associated factor X; Translin-associated protein X; translin-like protein; Trax; TSNAX
Gene Symbols TSNAX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSNAX. Recombinant protein control fragment (Product #RP-94100).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.