Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tsp23 Monoclonal Antibody (tsp232A), QED Bioscience™
SDP

Catalog No. 502277416
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7416 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7416 Supplier Leinco Technologies Inc Supplier No. 11137100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

tsp23 Monoclonal antibody specifically detects tsp23 in Human, Mouse samples. It is validated for Immunohistochemistry (PFA fixed), Western Blot

Specifications

Antigen tsp23
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone tsp232A
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene PTGES3
Gene Accession No. Q15185, Q9R0Q7
Gene Alias p23 homolog; Transcript similar p23
Host Species Mouse
Immunogen Recombinant tsp23
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10728, 56351
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.