Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSP50 Monoclonal Antibody (2D8)
GREENER_CHOICE

Catalog No. PIMA521771
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521771 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521771 Supplier Invitrogen™ Supplier No. MA521771
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: QELKYSNYVR PICLPGTDYV LKDHSRCTVT GWGLSKADGM WPQFRTIQEK EVIILNNKEC DNFYHNFTKI PTLVQIIKSQ MMCAEDTHRE KFCYELTGEP.

Testes-specific protease 50 (TSP50) is a gene located within the chromosome region 3p21.31, primarily expressed in testicular tissue, and is homologous to serine proteases. However, TSP50 contains a distinctive catalytic triad where serine, a crucial catalytic residue, is replaced by threonine, allowing it to exhibit a novel protease activity. TSP50 is recognized for its role as a pro-oncogene, contributing significantly to cancer progression by promoting aerobic glycolysis, known as the Warburg effect, in hepatocellular carcinoma cells through interaction with pyruvate kinase M2 isoform (PKM2). This function underlines its involvement in cancer metabolism, as it attenuates colitis by regulating TGF-β signaling pathways. Furthermore, studies have linked TSP50 to cancer stem cell maintenance, driving stem-like properties in breast cancer cells, thereby facilitating tumor initiation and self-renewal capabilities. These characteristics highlight TSP50's potential as a target in therapeutic interventions for cancer treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSP50
Applications ELISA
Classification Monoclonal
Clone 2D8
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PRSS50
Gene Accession No. Q9UI38
Gene Alias 79H19C; Cancer/testis antigen 20; CT20; Probable threonine protease PRSS50; protease, serine 50; protease, serine, 50; PRSS50; Serine protease 50; testes specific protease 50; testes-specific protease 50; testicular tissue protein Li 210; testis-specific protease-like protein 50; Tsp50
Gene Symbols PRSS50
Host Species Mouse
Immunogen PRSS50 (NP_037402.1, 214 a.a. ~ 313 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29122
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.