Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSP50 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501728079 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8079 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8079 Supplier Proteintech Group Inc Supplier No. 125741AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in proteolysis through its threonine endopeptidase activity.

Specifications

Antigen TSP50
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRSS50
Gene Accession No. Q8BLH5, Q9UI38
Gene Alias Cancer/testis antigen 20, CT20, PRSS50, Serine protease 50, testes protease 50, TSP50
Gene Symbols PRSS50
Host Species Rabbit
Immunogen TSP50 Fusion Protein Ag3247
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235631, 29122
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.