Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSP50 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987466
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7466 Supplier Bioss Supplier No. BS6745R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in proteolysis through its threonine endopeptidase activity.

Specifications

Antigen TSP50
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PRSS50
Gene Accession No. Q9UI38
Gene Alias 79H19C; Cancer/testis antigen 20; CT20; Probable threonine protease PRSS50; protease, serine 50; protease, serine, 50; PRSS50; Serine protease 50; testes specific protease 50; testes-specific protease 50; testicular tissue protein Li 210; Testis-specific protease-like protein 50; Tsp50
Gene Symbols PRSS50
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TSP50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29122
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.