Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tspan-33, Mouse anti-Mouse, Human, FITC, Clone: 29A6.2, Millipore Sigma™
Mouse Monoclonal Antibody
$192.34 - $482.78
Specifications
Antigen | Tspan-33 |
---|---|
Clone | 29A6.2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT911F25
|
MilliporeSigma
MABT911F25UL |
25 μL |
Each for $192.34
|
|
|||||
MABT911F100
|
MilliporeSigma
MABT911F100UL |
100 μL |
Each for $482.78
|
|
|||||
Description
Anti-Tspan-33, clone 29A6.2, FITC conjugate, Cat. No. MABT911F, is a mouse monoclonal antibody that detects Tetraspanin-33 and has been tested for use in Flow Cytometry.
Tetraspanin-33 (UniProt: Q8R3S2; also known as Tspan-33, Penumbra, mPen, Proerythroblast new membrane) is encoded by the Tspan33 (also known as Pen) gene (Gene ID: 232670) in murine species. Tetraspanins are evolutionarily conserved proteins and thus far 33 of them have been described in human and murine species. Tspan-33 is a multi-pass, disulfide-linked homodimeric membrane protein of the tetraspanin (TM4SF) family. It has been reported that Tspan-33 is highly expressed in activated B cells. Its expression has also been reported in malignant B cells. Two isoforms of Tspan-33 have been described that are produced by alternative splicing. Tspan-33 plays an important role in normal erythropoiesis and in the differentiation of erythroid progenitors. Tspan-33 contains four transmembrane domains (aa 25-45, 65-85, 97-117, 236-256), three cytoplasmic domains (aa 1-24, 86-96, 257-283), and two extracellular domains (aa 46-64, 118-235). Several tetraspanins interact with specific membrane proteins via their second extracellular domain (aa 118-235) and incorporate them into tetraspanin-enriched microdomains to modulate the effectiveness of signal transduction by those membrane proteins. (Ref.: Luu, VP., et al. (2013). Clin. Immunol. 149(3): 388-399).Specifications
Tspan-33 | |
Flow Cytometry | |
FITC | |
Mouse | |
Cell Structure | |
Purified mouse monoclonal antibody IgM in PBS with 0.1% sodium azide, with 1.5% BSA. | |
NP_666285.1 | |
BaF3 cells expressing full-length murine Tspan-33 fused with a Myc-His tag at the C-terminus. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
29A6.2 | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Human | |
Tetraspanin-33;Penumbra;mPen;Proerythroblast new membrane | |
Tspan33;Pen | |
IgM κ | |
Protein G purified | |
Clone 29A6.2 specifically detects Tspan-33 and targets an epitope with in the extracellular domain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title