Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN1 Antibody (4/12), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP312060MFV450
Description
TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
TSPAN1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
10103 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunocytochemistry | |
2024-04-12 00:00:00 | |
ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 | |
This antibody was raised by immunising BALB/c mice with the major extracellular domain of tetraspanin 1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction