Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121521
This item is not returnable.
View return policy
Description
TSPAN1 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications
TSPAN1 | |
Polyclonal | |
Unconjugated | |
TSPAN1 | |
9030418M05Rik; NET1; tetraspan 1; Tetraspan NET-1; tetraspanin 1; Tetraspanin TM4-C; Tetraspanin-1; TM4C; TM4SF; TSPAN1; Tspan-1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10103, 66805 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O60635, Q99J59 | |
TSPAN1 | |
Recombinant protein Tetraspanin 1. The antigen corresponds to amino acid range 112-215 of the target protein. | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction