Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566867 Supplier Invitrogen™ Supplier No. PA566867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSDELKQHLN RTLAENYGQP GATQITASVD RLQQDFKCCG SN Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

TSPAN11 gene ontology annotations related to this gene include cell surface receptor signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSPAN11
Gene Accession No. A1L157
Gene Alias 1110014F12Rik; AU067806; tetraspanin 11; tetraspanin-11; TSPAN11; Tspan-11; VSSW1971
Gene Symbols TSPAN11
Host Species Rabbit
Immunogen Recombinant Human TSPAN11. Recombinant protein control fragment (Product #RP-105406).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.