Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN12 Antibody (921938), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8910H
Description
TSPAN12 Monoclonal antibody specifically detects TSPAN12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| TSPAN12 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 921938 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, NET-2, ten-eleven translocation-1, tetraspanin-12 | |
| NS0 mouse myeloma cell line transfected with human TSPAN12, Met1-Leu305 | |
| RUO | |
| 23554 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction