Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563538 Supplier Invitrogen™ Supplier No. PA563538
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMVPVQWSDM VTLKARMTNY GLPRYRWLTH AWNFFQREFK CCGVVYFTDW LEMTEMDWPP DSCCVREFPG CSKQAHQEDL SDLYQEGCGK KMY Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

TSPAN12 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSPAN12
Gene Accession No. O95859
Gene Alias 9030619E17; AI426782; AI663988; AW111457; EVR5; MGC137611 protein; NET2; NET-2; Tetraspan NET-2; tetraspanin 12; tetraspanin-12; Tm4sf12; transmembrane 4 superfamily member 12; TSPAN12; Tspan-12; UNQ774/PRO1568; zgc:63880
Gene Symbols TSPAN12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSPAN12. Recombinant protein control fragment (Product #RP-104048).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23554
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.