Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533092
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533092 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533092 Supplier Invitrogen™ Supplier No. PA533092
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Elephant, Panda, Horse, Rabbit (100%).

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TSPAN13
Gene Accession No. O95857, Q5FVL6, Q9D8C2
Gene Alias 1100001I23Rik; NET6; NET-6; tetraspan NET-6; Tetraspan Tspan-13; tetraspanin 13; tetraspanin-13; Tm4sf13; transmembrane 4 superfamily member 13; transmembrane 4 superfamily member tetraspan NET-6; Tspan13; tspan-13; UNQ260/PRO296
Gene Symbols TSPAN13
Host Species Rabbit
Immunogen Synthetic 13 amino acid peptide from internal region of human TSPAN13.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100066103, 100358391, 27075, 366602, 482332, 66109
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.