Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552434
Description
Immunogen sequence: ACLALNQEQQ GQLLEVGWNN TASARNDIQR NLNCCGFRSV NPNDTCLASC VKSDHSCSPC APIIGEYAGE V Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
Specifications
TSPAN13 | |
Polyclonal | |
Unconjugated | |
TSPAN13 | |
1100001I23Rik; NET6; NET-6; tetraspan NET-6; Tetraspan Tspan-13; tetraspanin 13; tetraspanin-13; Tm4sf13; transmembrane 4 superfamily member 13; transmembrane 4 superfamily member tetraspan NET-6; Tspan13; tspan-13; UNQ260/PRO296 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27075 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95857 | |
TSPAN13 | |
Recombinant protein corresponding to Human TSPAN13. Recombinant protein control fragment (Product #RP-88669). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction