Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN16 Monoclonal Antibody (5A2C10)
GREENER_CHOICE

Catalog No. FERMA548725
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548725 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548725 Supplier Invitrogen™ Supplier No. MA548725
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSPAN16 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN16
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5A2C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TSPAN16
Gene Accession No. Q9UKR8
Gene Alias tetraspanin 16; tetraspanin TM4-B; Tetraspanin-16; TM4-B; TM4SF16; TM-8; transmembrane 4 superfamily member 16; TSPAN16; Tspan-16
Gene Symbols TSPAN16
Host Species Mouse
Immunogen Purified recombinant fragment of human TSPAN16 (AA: 116-245) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26526
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.