Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569207
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Regulates ADAM10 maturation.
Specifications
TSPAN17 | |
Polyclonal | |
Unconjugated | |
Tspan17 | |
2210021G21Rik; AI047581; F-box only protein 23; F-box only protein 23, transmembrane 4 superfamily member 17; FBX23; Fbxo23; fc49c03; gamma-hydroxybutyrate receptor; GHB-R; tetraspan protein SB134; tetraspanin 17; tetraspanin-17; TM4SF17; transmembrane 4 superfamily member 17; transmembrane 4 superfamily member 17 isoform b; TSPAN17; tspan-17; wu:fc49c03; zgc:158284 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26262 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96FV3 | |
Tspan17 | |
synthetic peptide directed towards the N terminal region of human TSPAN17. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction