Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565066 Supplier Invitrogen™ Supplier No. PA565066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NDWNLNIYFNC TDLNPSRERC GVPFSCCVRD PAEDVLNTQC GYDVRLKLEL EQQGFIHTKG CVGQFEKWL Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

Regulates ADAM10 maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN17
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tspan17
Gene Accession No. Q96FV3
Gene Alias 2210021G21Rik; AI047581; F-box only protein 23; F-box only protein 23, transmembrane 4 superfamily member 17; FBX23; Fbxo23; fc49c03; gamma-hydroxybutyrate receptor; GHB-R; tetraspan protein SB134; tetraspanin 17; tetraspanin-17; TM4SF17; transmembrane 4 superfamily member 17; transmembrane 4 superfamily member 17 isoform b; TSPAN17; tspan-17; wu:fc49c03; zgc:158284
Gene Symbols Tspan17
Host Species Rabbit
Immunogen Recombinant Human TSPAN17. Recombinant protein control fragment (Product #RP-105829).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26262
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.