Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN2 Antibody (822509) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7876J
Description
TSPAN2 Monoclonal antibody specifically detects TSPAN2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
TSPAN2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10100 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
822509 | |
Flow Cytometry, CyTOF-ready | |
FLJ12082, NET3, new EST tetraspan 3, tetraspan 2, Tetraspan NET-3, tetraspan TM4SF, tetraspanin 2, tetraspanin 2 isoform, tetraspanin-2, TSN2, TSPAN-2 | |
NS0 mouse myeloma cell line transfected with human TSPAN2, Met1-Ile221, Accession # O60636 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction