Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107043 Supplier Invitrogen™ Supplier No. PA5107043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 79%; Dog: 79%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 86%; Rat: 93%.

TSPAN2 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN2
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene TSPAN2
Gene Accession No. O60636
Gene Alias 6330415F13Rik; B230119D02Rik; NET3; new EST tetraspan 3; tetraspan 2; tetraspan NET-3; tetraspan TM4SF; tetraspanin 2; tetraspanin 2 isoform; tetraspanin-2; TSN2; TSPAN2; TSPAN-2
Gene Symbols TSPAN2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TSPAN2 (aa 108-158).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10100
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.