Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528514
Description
Recommended positive controls: 293T, A431, HeLa, mouse kidney. Predicted reactivity: Mouse (84%), Rat (83%), Pig (84%), Rhesus Monkey (86%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. Two alternative transcripts encoding different isoforms have been described.
Specifications
TSPAN3 | |
Polyclonal | |
Unconjugated | |
TSPAN3 | |
1700055K04Rik; H41D4 protein; OAP-1; OSP-associated protein 1; tetraspan 3; tetraspan TM4SF; tetraspanin 3; tetraspanin TM4-A; tetraspanin TM4-A homolog; tetraspanin-3; TM4-A; Tm4sf8; Transmembrane 4 superfamily member 8; TSPAN3; TSPAN-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10099, 56434 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O60637, Q9QY33 | |
TSPAN3 | |
Recombinant fragment contain a sequence corresponding to a region within amino acids 1 and 233 of TSPAN3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction