Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589708
Description
Immunogen sequence: SCLAINRSKQ TDVINASWWV MSNKTRDELE RSFDCCGLFN LTTLYQQDYD FCTAICKSQS PTCQMCGEKF LKHSDEALKI L; Positive Samples: Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Rat ovary; Cellular Location: Membrane, Multi-pass membrane protein.
TSPAN31 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is thought to be involved in growth-related cellular processes. It is associated with tumorigenesis and osteosarcoma.
Specifications
TSPAN31 | |
Polyclonal | |
Unconjugated | |
TSPAN31 | |
2700085A14Rik; AW742554; sarcoma amplified sequence; sarcoma-amplified sequence; Sarcoma-amplified sequence homolog; SAS; tetraspanin 31; tetraspanin-31; transmembrane 4 protein; transmembrane protein; TSPAN31; Tspan-31; Unknown (protein for MGC:133819) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
362890, 6302, 67125 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.80 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q12999, Q5U1V9, Q9CQ88 | |
TSPAN31 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 95-175 of human TSPAN31 (NP_0059721). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction