Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542904
Description
Peptide sequence: YEQAMKGTSH VRRQELAAIQ DVFLCCGKKS PFSRLGSTEA DLCQGEEAAR Sequence homology: Human: 100%.
This gene, which is a member of the tetraspanin superfamily, is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of chromosome 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian and breast cancers. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4, escapes imprinting. This gene may play a role in malignancies and diseases that involve this region, and it is also involved in hematopoietic cell function. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
TSPAN32 | |
Polyclonal | |
Unconjugated | |
TSPAN32 | |
AML1-regulated transmembrane protein 1; ART1; Art-1; AW208513; BB235973; D7Wsu37e; FLJ17158; FLJ97586; MGC22455; pan hematopoietic expression; pan-hematopoietic expression protein; Phemx; PHMX; protein Phemx; tetraspanin; tetraspanin 32; tetraspanin-32; TSPAN32; Tspan-32; Tssc6; tumor-suppressing STF cDNA 6; tumor-suppressing subchromosomal transferable fragment 6; tumor-suppressing subchromosomal transferable fragment cDNA 6; tumor-suppressing subtransferable candidate 6 | |
Rabbit | |
Protein A | |
RUO | |
10077 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96QS1 | |
TSPAN32 | |
Synthetic peptide directed towards the middle region of human TSPAN32. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction