Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN33 Monoclonal Antibody (8A2A9)
GREENER_CHOICE

Catalog No. FERMA548820
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548820 Supplier Invitrogen™ Supplier No. MA548820
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSPAN33 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN33
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8A2A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TSPAN33
Gene Accession No. Q86UF1
Gene Alias 1300010A20Rik; hPen; hypothetical protein LOC539284; mPen; PEN; PEN.; Penumbra; proerythroblast new membrane; RGD1560915; tetraspanin 33; tetraspanin-33; Tetraspanin-33-like protein; Tspan33; tspan-33
Gene Symbols TSPAN33
Host Species Mouse
Immunogen Purified recombinant fragment of human TSPAN33 (AA:Extra(46-64)+Extra(118-235) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340348
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.