Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526404
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526404 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526404 Supplier Invitrogen™ Supplier No. PA526404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine and mouse based on sequence homology.

TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN33
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TSPAN33
Gene Accession No. Q86UF1
Gene Alias 1300010A20Rik; hPen; hypothetical protein LOC539284; mPen; PEN; PEN.; Penumbra; proerythroblast new membrane; RGD1560915; tetraspanin 33; tetraspanin-33; Tetraspanin-33-like protein; Tspan33; tspan-33
Gene Symbols TSPAN33
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 254-283 amino acids from the C-terminal region of human TSPAN33.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340348
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.