Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN33 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554186
Description
Immunogen sequence: NAIVHYRDDL DLQNLIDFGQ KKFSCCGGIS YKDWSQNMYF NCSEDNPSRE RCSVPYSCCL PTPDQAVINT MCGQGMQAFD YLEASKVIYT NGCIDKLVNW IHSNLFL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.
Specifications
TSPAN33 | |
Polyclonal | |
Unconjugated | |
TSPAN33 | |
1300010A20Rik; hPen; hypothetical protein LOC539284; mPen; PEN; PEN.; Penumbra; proerythroblast new membrane; RGD1560915; tetraspanin 33; tetraspanin-33; Tetraspanin-33-like protein; Tspan33; tspan-33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UF1 | |
TSPAN33 | |
Recombinant protein corresponding to Human TSPAN33. Recombinant protein control fragment (Product #RP-91166). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction