Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN33 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592356
Description
Immunogen sequence: NTMCGQGMQA FDYLEASKVI YTNGCIDKLV NWIHSNLFLL GGVALGLAIP QLVGILLSQI LVNQIKDQIK LQLYNQQHRA DPWY; Positive Samples: Mouse brain, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.
TSPAN33 plays an important role in normal erythropoiesis. It has a role in the differentiation of erythroid progenitors.
Specifications
TSPAN33 | |
Polyclonal | |
Unconjugated | |
TSPAN33 | |
1300010A20Rik; hPen; hypothetical protein LOC539284; mPen; PEN; PEN.; Penumbra; proerythroblast new membrane; RGD1560915; tetraspanin 33; tetraspanin-33; Tetraspanin-33-like protein; Tspan33; tspan-33 | |
Rabbit | |
Affinity chromatography | |
RUO | |
232670, 340348, 500065 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.85 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q86UF1, Q8R3S2 | |
TSPAN33 | |
A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human TSPAN33 (NP_8486571). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction