Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542906 Supplier Thermo Scientific Supplier No. PA542906
Add to Cart
Add to Cart

TSPAN5 Polyclonal Antibody

TSPAN5 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN5
Applications Immunohistochemistry, Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TSPAN5
Gene Accession No. P62079
Gene Alias 2810455A09Rik; 4930505M03Rik; NET4; NET-4; tetraspan 5; tetraspan NET-4; tetraspan TM4SF; tetraspanin 5; tetraspanin Tspan-5; tetraspanin-5; TM4SF9; transmembrane 4 superfamily member 9; transmembrane 4 superfamily, member 8; Tspan5; tspan-5
Gene Symbols TSPAN5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TSPAN5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10098
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.