Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598218
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598218 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598218 Supplier Invitrogen™ Supplier No. PA598218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSPAN5 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

TSPAN5 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TSPAN5
Gene Accession No. P62079
Gene Alias 2810455A09Rik; 4930505M03Rik; NET4; NET-4; tetraspan 5; tetraspan NET-4; tetraspan TM4SF; tetraspanin 5; tetraspanin Tspan-5; tetraspanin-5; TM4SF9; transmembrane 4 superfamily member 9; transmembrane 4 superfamily, member 8; Tspan5; tspan-5
Gene Symbols TSPAN5
Host Species Rabbit
Immunogen Recombinant Human Tetraspanin-5 protein (114-232AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10098
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.