Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533093
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533093 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533093 Supplier Invitrogen™ Supplier No. PA533093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Rabbit (100%) Elephant, Panda, Horse, Pig (95%) Bovine, Bat (89%).

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and is highly similar in sequence to the transmembrane 4 superfamily member 2. The use of alternate polyadenylation sites has been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TSPAN6
Gene Accession No. O43657
Gene Alias 6720473L21Rik; A15 homolog; AI316786; Putative NF-kappa-B-activating protein 321; T245; T245 protein; tetraspan TM4SF; tetraspanin 6; tetraspanin TM4-D; Tetraspanin-6; Tm4sf; TM4SF6; Transmembrane 4 superfamily member 6; TSPAN6; Tspan-6; UNQ767/PRO1560
Gene Symbols TSPAN6
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from internal region of human TSPAN6.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100344358, 7105
Target Species Human, Rabbit, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.