Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPAN7 Monoclonal Antibody (01)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530976
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TALLA-1/TSPAN7/CD231.
TSPAN7 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and may have a role in the control of neurite outgrowth. It is known to complex with integrins. This gene is associated with X-linked mental retardation and neuropsychiatric diseases such as Huntington's chorea, fragile X syndrome and myotonic dystrophy.
Specifications
TSPAN7 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P41732 | |
TSPAN7 | |
Recombinant Human TALLA-1/TSPAN7/CD231 Protein (Arg113-Met213). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
01 | |
Unconjugated | |
TSPAN7 | |
1200014P11Rik; A15; AI323365; CCG-B7; CD231; CD231 antigen; cell surface glycoprotein A15; DXS1692E; membrane component chromosome X surface marker 1; membrane component, X chromosome, surface marker 1; MRX58; Mxs1; neuroprotective protein 5; PE31; R74651; TALLA; TALLA homolog; TALLA-1; T-cell acute lymphoblastic leukemia associated antigen 1; T-cell acute lymphoblastic leukemia-associated antigen 1; tetraspanin 7; tetraspanin protein; tetraspanin-7; TM4SF2; TM4SF2b; transmembrane 4 superfamily 2b; transmembrane 4 superfamily member 2; Transmembrane 4, superfamily member 2 (cell surface glycoprotein A15); transmembrane protein A15; TSPAN7; tspan-7 | |
Mouse | |
Protein A | |
RUO | |
7102 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction